missing translation for 'onlineSavingsMsg'
Learn More

CD166 Mouse anti-Human, PE, Clone: 3A6, BD

Mouse Monoclonal Antibody

£71.63 - £267.41

Specifications

Antigen CD166
Clone 3A6
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15870579
View Documents
BD Biosciences
560903
25 Tests
ONLINE EXCLUSIVE PRICE
£74.90
£71.63 / Each
Save £3.27
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15899688
View Documents
BD Biosciences
559263
100 Tests
ONLINE EXCLUSIVE PRICE
£279.00
£267.41 / Each
Save £11.59
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 3A6 monoclonal antibody specifically binds to the activated leukocyte cell adhesion molecule (ALCAM), a 100-105 kDa transmembrane glycoprotein, also known as the CD6 ligand, CD166. It is composed of an immunoglobulin-like extracellular domain, a transmembrane region and a short cytoplasmic tail. ALCAM belongs to the Ig superfamily of proteins and is expressed on neurons, activated T cells, activated monocytes, epithelial cells and fibroblasts. CD166 plays an important role in mediating adhesion interactions between thymic epithelial cells and CD6+ cells during intrathymic T-cell development.

Specifications

Specifications

CD166
Flow Cytometry
PE
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
3A6
Monoclonal
Mouse
Human
IgG1 κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.