missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD166 Monoclonal Antibody (200622)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA523930
This item is not returnable.
View return policy
Description
In Western blots, shows 100% cross-reactivity with recombinant human (rh) ALCAM and no cross-reactivity with rhBCAM, rhEpCAM, recombinant mouse (rm) MAdCAM-1, rhMCAM, rmNCAM-L1, rmOCAM, or rmTROP-2. Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes activated leukocyte cell adhesion molecule (ALCAM), also known as CD166 (cluster of differentiation 166), which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. This protein binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
Specifications
| CD166 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q61490 | |
| ALCAM | |
| Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166 Trp28-Lys527. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Western Blot | |
| 200622 | |
| Unconjugated | |
| ALCAM | |
| activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
| Rat | |
| Protein A/G | |
| RUO | |
| 11658 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction