Learn More
Invitrogen™ CD166 (ALCAM) Monoclonal Antibody (eBioALC48), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ 14-1661-37
Description
Activated leukocyte cell adhesion molecule (ALCAM; CD166) is a 105 kD cell-surface adhesion molecule belonging to the Ig superfamily of receptors. ALCAM is a type I transmembrane glycoprotein, possessing five extracellular Ig domains, a transmembrane domain, and a short cytoplasmic tail. ALCAM is involved in homotypic adhesion, as well as heterotypic adhesion with its natural ligand, CD6. A soluble isoform of ALCAM has been shown to inhibit homophilic adhesive interactions. ALCAM is expressed on a variety of cell types, including activated T and B cells, monocyte-derived dendritic cells, and thymic epithelial cells. ALCAM expression has also been used as a marker for the isolation of pluripotent stem cells.
This gene encodes activated leukocyte cell adhesion molecule (ALCAM), also known as CD166 (cluster of differentiation 166), which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. This protein binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
Specifications
| CD166 (ALCAM) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q61490 | |
| ALCAM | |
| Affinity chromatography | |
| RUO | |
| 11658 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| eBioALC48 | |
| Unconjugated | |
| ALCAM | |
| activated leukocyte cell adhesion molecule; AI853494; ALCAM; BEN; CD166; CD166 antigen; cell adhesion molecule; DM-GRASP; FLJ38514; HB2; KG-CAM; MEMD; MGC71733; MuSC; Protein DM-GRASP; Protein MEMD; SB-10 antigen; SC1; unnamed protein product | |
| Rat | |
| 2 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.