missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD163L1 Antibody (SR2344), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | CD163L1 |
|---|---|
| Clone | SR2344 |
| Dilution | Immunohistochemistry 1:50-1:200 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041590
|
Novus Biologicals™
NBP3-27697-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041562
|
Novus Biologicals™
NBP3-27697-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD163L1 Monoclonal antibody specifically detects CD163L1 in Human samples. It is validated for ImmunohistochemistrySpecifications
| CD163L1 | |
| Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD163 antigen B, CD163 antigen-like 1, CD163 molecule-like 1, CD163B, M160, scavenger receptor cysteine-rich type 1 protein M160, UNQ6434/PRO23202 | |
| A synthesized peptide derived from human CD163L1 (Uniprot #: Q9NR16) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR2344 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| CD Markers, Cell Biology, Immunology | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 283316 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title