missing translation for 'onlineSavingsMsg'
Learn More

CD163L1 Antibody (CD163L1/7972), DyLight 550, Novus Biologicals™

Product Code. 18565909 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18565909 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18565909 Supplier Novus Biologicals Supplier No. NBP324185R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD163L1 Monoclonal antibody specifically detects CD163L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD163L1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CD163L1/7972
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias CD163 antigen B, CD163 antigen-like 1, CD163 molecule-like 1, CD163B, M160, scavenger receptor cysteine-rich type 1 protein M160, UNQ6434/PRO23202
Host Species Mouse
Immunogen Recombinant fragment of human CD163L1 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 283316
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.