missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163L1 Antibody (CD163L1/7971) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24128JF585
This item is not returnable.
View return policy
Description
CD163L1 Monoclonal antibody specifically detects CD163L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD163L1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD163 antigen B, CD163 antigen-like 1, CD163 molecule-like 1, CD163B, M160, scavenger receptor cysteine-rich type 1 protein M160, UNQ6434/PRO23202 | |
| Recombinant fragment of human CD163L1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| CD Markers, Cell Biology, Immunology | |
| 283316 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CD163L1/7971 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction