missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD163 Recombinant Rabbit Monoclonal Antibody (23GB3750), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CD163 |
|---|---|
| Clone | 23GB3750 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087883
![]() |
Invitrogen™
MA554106 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089799
|
Invitrogen™
MA554107 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD163 Recombinant Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CD163 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q86VB7 | |
| 9332 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3750 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163 | |
| A synthetic peptide of human CD163 (1001-1156AA). | |
| Primary | |
| -20°C | |
| CD163 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title