missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Rabbit anti-Human, Clone: M130/3708R, Novus Biologicals™
Rabbit Monoclonal Antibody
£431.00
Specifications
| Antigen | CD163 |
|---|---|
| Clone | M130/3708R |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
| CD163 | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| 9332 | |
| IgG κ | |
| Protein A or G purified |
| M130/3708R | |
| ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| Synthetic peptide corresponding to CD163 residues within aa1056-1156 of CD163 was used as an immunogen. | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title