missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD163 Monoclonal Antibody (MAC 2-158), NovaFluor™ Red 685, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£151.00 - £336.00
Specifications
| Antigen | CD163 |
|---|---|
| Clone | MAC 2-158 |
| Concentration | 0.2 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30245286
|
Invitrogen™
H088T02R02-A |
25 Tests |
£151.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30245421
|
Invitrogen™
H088T03R02-A |
100 Tests |
£336.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow CytometrySpecifications
| CD163 | |
| 0.2 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| MAC 2-158 | |
| Flow Cytometry | |
| NovaFluor Red 685 | |
| Mouse | |
| Human | |
| Q86VB7 | |
| 9332 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD163 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title