missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD163 Antibody (OR2805), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28038-50ug
This item is not returnable.
View return policy
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD163 | |
| Monoclonal | |
| Unconjugated | |
| Q86VB7 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| OR2805 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| CD163 | |
| 50 μg | |
| Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity | |
| 9332 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction