missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Antibody (CL10658) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21179AF488
This item is not returnable.
View return policy
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| CD163 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| This antibody was developed using a recombinant protein derived from Q86VB7, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity | |
| 9332 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10658 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction