missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD162 Recombinant Rabbit Monoclonal Antibody (001), FITC
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£32.50 - £82.40

Specifications

Antigen CD162
Clone 001
Concentration 0.1 mg/mL
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30534826
GREENER_CHOICE
View Documents
Invitrogen™
MA560485
100 Tests
£82.40
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30533789
View Documents
Invitrogen™
MA560486
25 Tests
£32.50
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD162 Recombinant Monoclonal Antibody for Flow

CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.
Specifications

Specifications

CD162
0.1 mg/mL
Flow Cytometry
FITC
Rabbit
RUO
PBS with 0.5% BSA and 0.09% sodium azide
Q62170
20345
Recombinant Mouse PSGL-1/CD162 protein (Met1-Cys307).
Antibody
001
4°C, store in dark, DO NOT FREEZE!
Recombinant Monoclonal
Liquid
IgG
Mouse
SELPLG
CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG
SELPLG
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.