missing translation for 'onlineSavingsMsg'
Learn More

CD162 Rat anti-Mouse, BUV615, Clone: 2PH1, BD OptiBuild™

Rat Monoclonal Antibody

Brand:  BD Biosciences 752333

Product Code. 17253025

  • ONLINE EXCLUSIVE PRICE
    £491.00
    £470.80 / 50µg
    Save £20.20
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD162 Monoclonal antibody specifically detects CD162 in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD162
Monoclonal
0.2 mg/mL
Ms CD162 BUV615 2PH1 50 μg
Ovalbumin-conjugated peptide covering amino acids 42 to 60 of mouse PSGL-1
RUO
CD162
Refrigerate (2 to 8°C)
Mouse
Flow Cytometry
2PH1
Brilliant Ultraviolet 615
Rat
1 x 50 μg
Primary
Mouse
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.