missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD162 (PSGL-1) Monoclonal Antibody (4RA10), PE-eFluor™ 610, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£166.00 - £418.00
Specifications
| Antigen | CD162 (PSGL-1) |
|---|---|
| Clone | 4RA10 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.Specifications
| CD162 (PSGL-1) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 20345 | |
| Mouse PSGL-1 human IgG1 fusion protein | |
| Primary | |
| Antibody |
| 4RA10 | |
| Flow Cytometry | |
| PE-eFluor 610 | |
| Rat | |
| Mouse | |
| Q62170 | |
| SELPLG | |
| IgG1 κ | |
| Affinity chromatography | |
| SELPLG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title