Learn More
CD161f Rat anti-Mouse, Brilliant Violet 421, Clone: C8, BD
Rat Monoclonal Antibody
Brand:  BD Biosciences 566304
Product Code. 15844929
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The C8 monoclonal antibody specifically recognizes C57BL/6 mouse CD161 antigen-like family member F (CD161f ). CD161f is also known as Natural killer cell surface protein NKR-P1F (Nkrp1f), and is encoded by Klrb1f /(Killer cell lectin-like receptor subfamily B member 1F). CD161f is expressed on natural killer (NK) cells, and some dendritic cells and activated lymphocytes. CD161f binds to C-type lectin related protein c, d, and g (Clr-c, d, g) which are expressed on several cell types including dendritic cells, B cells, and activated T cells. CD161f-mediated signaling can reportedly costimulate T cell proliferation and IL-2 production, and may play a role in dendritic cell maturation.
The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD161f
C8
Brilliant Violet 421
Klrb1f, Nkrp1-f, Nkrp1f, NKR-P1F
Mouse NKRP1F Transfected Cell Line
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
Affinity Purified
RUO
Mouse
IgG1 κ