missing translation for 'onlineSavingsMsg'
Learn More

CD161a Mouse anti-Rat, FITC, Clone: 10/78, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561781

 View more versions of this product

Product Code. 15861329

  • ONLINE EXCLUSIVE PRICE
    £139.00
    £132.86 / Each
    Save £6.14
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 10/78 antibody reacts with NKR-P1A, a 60-kDa homodimer expressed on all natural killer (NK) cells and a small subset of T lymphocytes. The 10/78 antibody competes with the previously described 3.2.3 antibody for binding to the antigen. NKR-P1A is a type-II integral membrane protein with an extracellular C-type lectin domain, which is an NK cell-activating receptor specific for tumor target cells. Many rat dendritic cells have been shown to express NKR-P1A, and a subpopulation of these cells has cytotoxic activity. NKR-P1A has also been detected at low levels on peripheral blood monocytes, and its expression is upregulated in IFN-γ-activated monocytes, specifically in a subpopulation of large monocytes with phagocytic capacity. Furthermore, activated peripheral blood neutrophils may express a low level of NKR-P1A. In the mouse and rat, three members of the NKR-P1 gene family have been identified; but in the human gene family, a single NKR-P1 homologue has been discovered and designated Cd161

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD161a
Monoclonal
0.5mg/mL
NKR-P1A
Mouse
Affinity Purified
RUO
Rat
IgG1 κ
Flow Cytometry
Oct-78
FITC
Aqueous buffered solution containing ≤0.09% sodium azide.
Not reported
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.