missing translation for 'onlineSavingsMsg'
Learn More

CD161/NK1.1 Mouse anti-Mouse, Clone: PK136, Novus Biologicals™

Product Code. 18322436 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322436 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322436 Supplier Novus Biologicals Supplier No. NBP30753220UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD161/NK1.1 Monoclonal specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD161/NK1.1
Applications Flow Cytometry, Immunofluorescence, Functional Assay
Classification Monoclonal
Clone PK136
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Functional
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
Host Species Mouse
Immunogen CD Mouse spleen and bone marrow cells enriched for NK-1+ cells
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 17059
Target Species Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.