Learn More
CD161/NK1.1 Antibody (PK-136) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-20165JF525
Product Code. 30171599
  • £343.00 / 0.10mL
Description
CD161/NK1.1 Monoclonal antibody specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry,ELISA,Cell depletion
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD161/NK1.1
Monoclonal
Janelia Fluor 525
CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
The original antibody was generated by immunizing (C3H X BALB)F1 mice with spleen cells enriched for NK-1 cells and bone marrow cells from CE mice.
0.1 mL
Innate Immunity, Signal Transduction
17059
Store at 4°C in the dark.
IgG κ
Flow Cytometry, ELISA, Depletion Assay
PK-136
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified