Learn More
CD161 Mouse, Qfluor 710, Clone: PK136, enQuireBio™
Mouse Monoclonal Antibody
Brand:  EnquireBio QAB78-QF710-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15962488
  • £89.95 / 25µg
Estimated Shipment: 11-07-2024
to see stock.
Specifications
CD161
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Klrb1c
Protein A/G Purified
RUO
17059
IgG2a κ
Flow Cytometry
PK136
Qfluor 710
NP_001153376
Mouse
25 μg
Primary
Store at 2-4°C.