Learn More
CD161 Mouse anti-Human, FITC, Clone: DX12, BD
Mouse Monoclonal Antibody
£54.57 - £462.00
Specifications
| Antigen | CD161 |
|---|---|
| Clone | DX12 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15825918
|
BD Biosciences
340536 |
50 Tests |
£462.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15808638
|
BD Biosciences
556080 |
100 Tests |
£55.55 £54.57 / Each Save £0.98 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DX12 monoclonal antibody specifically binds to human CD161 which is also known as Natural killer cell surface protein P1A (NKR-P1A). CD161 is encoded by the KLRB1 (Killer cell lectin-like receptor subfamily B member 1) gene. CD161 is a member of the C-type lectin superfamily and is also referred to as C-type lectin domain family 5 member B (CLEC5B). CD161 is a 80 kDa disulfide-linked homodimer, type II membrane glycoprotein. CD161 is expressed mostly on NK cell populations and on subsets of CD4+ and CD8+ αβ T cells, NKT cells and γδ T cells. Reports indicate that CD161 is expressed preferentially on CD45RO+ T cells, however, it can be found on a subset of thymocytes and fetal liver T cells. Its function has not been fully elucidated, but reports indicate that NKR-P1A may serve as a specific receptor for some NK cell targets. The DX12 antibody can reportedly inhibit spontaneous cytotoxicity mediated by certain NK cell clones.
Specifications
| CD161 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Human | |
| IgG1 κ |
| DX12 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Human NKR-P1A Transfected Mouse Fibroblasts | |
| Primary |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.