Learn More
Invitrogen™ CD160 Monoclonal Antibody (CNX46-3), FITC
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA517967
Product Code. 15344241
  • £308.00 / 100µg
Description
MA5-17967 targets CD160/BY55 antigen in FACS and WB applications and shows reactivity with Mouse samples. The MA5-17967 immunogen is mouse CD160.
CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. It is involved in cell proliferation, cell surface receptor signaling pathways, cellular defense response, and regulation of immune response.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD160
Monoclonal
0.1 mg/mL
PBS with 4-5mg/mL BSA and 0.02% sodium azide
O88875
CD160
Mouse CD160.
100 μg
Primary
Mouse
Antibody
IgG2a κ
Flow Cytometry, Western Blot
CNX46-3
FITC
CD160
AU045688; BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; natural killer cell receptor; natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598
Rat
Protein G
RUO
54215
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.