missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16/CD32 Rat, anti-Mouse, Unconjugated, Clone: 2.4G2, STEMCELL Technologies
Rat Monoclonal Antibody
£183.60 - £583.20
Specifications
| Antigen | CD16/CD32 |
|---|---|
| Clone | 2.4G2 |
| Applications | Immunoprecipitation, Flow Cytometry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17119221
|
Stemcell Technologies
60161.2 |
100 μg |
£183.60
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17109221
|
Stemcell Technologies
60161.1 |
500 μg |
£363.15
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199211
|
Stemcell Technologies
60161 |
1 mg |
£583.20
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The 2.4G2 antibody reacts with a shared epitope of Fc-gamma receptors CD16 (FcRIII) and CD32 (FcRII) in the mouse
- CD16 and CD32 are low affinity receptors for the Fc portion of IgG, with CD32 having a higher affinity for IgG aggregates and immune complexes
- CD16 and CD32 are expressed on B cells, monocytes, macrophages, NK cells, neutrophils, granulocytes, mast cells, and dendritic cells
- The binding of immunoglobulins to Fc receptors is involved in multiple immunological functions, including phagocytosis, cytotoxocity, and the modulation of immune responses
- The 2.4G2 antibody blocks the interaction between IgG and CD16 or CD32, and therefore is used as a pre-treatment to minimize non-specific binding of antibodies to FcR-bearing cells in applications such as flow cytometry, cell sorting, and immunofluorescence
Specifications
| CD16/CD32 | |
| Immunoprecipitation, Flow Cytometry, Immunofluorescence, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Primary |
| 2.4G2 | |
| Monoclonal | |
| Rat | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title