missing translation for 'onlineSavingsMsg'
Learn More

CD16/CD32 Monoclonal Antibody (93), Biotin, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0161-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229469

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Description: The 93 monoclonal antibody reacts with an epitope shared by mouse CD16 and CD32. CD16 (Fc gamma III Receptor) and CD32 (Fc gamma II Receptor) are the low affinity receptors for the mouse IgG Fc portion and are expressed by B cells, monocyte/macrophages, NK cells, and neutrophils. Applications Reported: The 93 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 93 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

CD16 is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by natural killer (NK) cells, and some T cells and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC (antibody dependent cell mediated cytotoxicity) by NK cells. IN comparison, CD32 is a 40 kD glycoprotein that acts as a low affinity receptor for IgG (also known as Fc gamma RII), and mediates several functions including endocytosis, activation of secretion, cytotoxicity and immunomodulation. CD32 is expressed by B cells, monocytes, granulocytes and platelets. Further, CD32 is involved in the phagocytosis of immune complexes, the regulation of antibody production by B-cells, and variations of the CD32 gene may increase vulnerability to systemic lupus erythematosus (SLE). Several transcript variants encoding different isoforms of CD32 have been found.
Specifications

Specifications

CD16/CD32
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P08101
Fcgr2b
Affinity chromatography
RUO
14130
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
93
Biotin
Fcgr2b
FCGR3, FCGR3A, FCGR3B, CD16A, CD16B, Fc gamma Receptor III, Fc gamma Receptor 3, RP11-5K23.1, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, FCGR2, CD32, Fc gamma Receptor II, CD32 receptor 2, Fc gamma Receptor 2, FCGR2A, FCGR2B, FCGR2C, Low affinity immunoglobulin gamma Fc region receptor II-b, CDw32, Fc-gamma RII-b, Fc-gamma-RIIb, FcRII-b, FCG2, Low affinity immunoglobulin gamma Fc region receptor II-c, Fc-gamma RII-c, Fc-gamma-RIIc, FcRII-c, FGFR2B
Rat
50 μg
Primary
Mouse
Antibody
IgG2a λ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.