Learn More
CD159c (NKG2C) Mouse anti-Human, Brilliant Violet 711, Clone: 134591, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 748164
This item is not returnable.
View return policy
Description
CD159c (NKG2C) Monoclonal antibody specifically detects CD159c (NKG2C) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD159c (NKG2C) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| CD159c (NKG2C) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 134591 | |
| Brilliant Violet 711 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |