Learn More
Invitrogen™ CD158f Monoclonal Antibody (UP-R1), PE, eBioscience™, Invitrogen™

Description
Description: This UP-R1 monoclonal antibody recognizes human CD158f, also known as KIR2DL5. CD158f is an inhibitory receptor and a member of the KIR (Killer cell Ig-like receptors) family of receptors, which recognizes subsets of HLA alleles. Expression is found mainly on a subset of NK cells with minimal expression also present on a minor subset of T lymphocytes in some individuals. This new member of the family is highly polymorphic and expression is not found on many human donors. Activation of the receptor and tyrosine phosphoryaltion recruits SHP-2 and SHP-1 to the ITIM cytoplasmic domain. The UP-R1 antibody has been shown to inhibit NK cytotoxicity. Applications Reported: This UP-R1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This UP-R1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Please note: There is significant donor variability observed. Only a small percentage of donors tested showed bright staining; all others were completely negative. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD158f |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | UP-R1 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | KIR2DL5A |
| Gene Accession No. | Q8N109 |
| Gene Alias | CD158; CD158F; CD158f1; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 5A; killer cell immunoglobulin-like receptor 2DL5A; killer cell immunoglobulin-like receptor KIR2DL5A; killer cell immunoglobulin-like receptor two domains long cytoplasmic tail 5; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A; killer-cell Ig-like receptor; KIR2DL; KIR2DL5; KIR2DL5.1; KIR2DL5.3; KIR2DL5A; NKAT; NKAT1 |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.