missing translation for 'onlineSavingsMsg'
Learn More

CD158a Mouse anti-Human, FITC, Clone: HP-3E4, BD

Mouse Monoclonal Antibody

£224.10 - £259.00

Specifications

Antigen CD158a
Clone HP-3E4
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15805918
View Documents
BD Biosciences
340531
50 Tests
£259.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15898618
View Documents
BD Biosciences
556062
100 Tests
ONLINE EXCLUSIVE PRICE
£234.00
£224.10 / Each
Save £9.90
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reacts wth a 58 kDa membrane glycoprotein, member of the killer-inhibitory receptor (KIR). The KIR family family consists of transmembrane glycoproteins of the Ig superfamily expressed on <1-8% of NK cells and a subset of T cells which are involved in recognition of MHC class I molecules on target cells and inhibit cytotoxicity. CD158a is an antigen which recognizes HLA-C molecules (-Cw0401 and -Cw1503). This antibody the purified format is suitable for staining acetone-fixed, frozen tissue sections. It was reported that this clone also cross-reacted with NKAT8 (CD158i) tranfectant cells.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

CD158a
Flow Cytometry
FITC
RUO
Human
HP-3E4
Monoclonal
Mouse
Immunology
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.