missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD155/PVR Rabbit anti-Human, Clone: 114, Novus Biologicals™
Rabbit Monoclonal Antibody
£299.00 - £392.00
Specifications
| Antigen | CD155/PVR |
|---|---|
| Clone | 114 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 ul/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261378
|
Novus Biologicals
NBP2-89272-50UL |
50 μg |
£299.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18204478
|
Novus Biologicals
NBP2-89272-100UL |
100 μg |
£392.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| CD155/PVR | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 ul/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
| Recombinant Human CD155/PVR protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 114 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| 5817 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title