missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD155/PVR Antibody (BLR074G), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-78711
This item is not returnable.
View return policy
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CD155/PVR | |
| Monoclonal | |
| Unconjugated | |
| Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA. with 0.09% Sodium Azide | |
| PVR | |
| A recombinant protein representing the extracellular domain of human PVR/CD155. | |
| 100 μL | |
| Primary | |
| Human | |
| Supernatant |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| BLR074G | |
| Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Virology Bacteria and Parasites | |
| 5817 | |
| Store at 2-8°C/ 1 year from date of receipt | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction