missing translation for 'onlineSavingsMsg'
Learn More

CD155/PVR Antibody (300903) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30735816 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735816 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735816 Supplier Novus Biologicals Supplier No. FAB2530I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD155/PVR Monoclonal antibody specifically detects CD155/PVR in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD155/PVR
Applications Western Blot
Classification Monoclonal
Clone 300903
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD155/PVR, Gly27-Asn343, Accession # ABM85069
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 5817
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.