Learn More
CD155 Monoclonal Antibody (D171), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513493
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-13493 targets CD155 in Western blot, FACS and ICC/IF applications and shows reactivity with Human, Mouse and Non-human primate samples. The MA5-13493 immunogen is HeLa cells.
The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
CD155 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P15151 | |
PVR | |
HeLa cells | |
500 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
D171 | |
Unconjugated | |
PVR | |
3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4 | |
Mouse | |
Protein G | |
RUO | |
52118, 5817 | |
4° C | |
Liquid |