missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD154 Mouse anti-Human, BV421, Clone: TRAP1, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566268
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
CD154 Monoclonal antibody specifically detects CD154 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Tekniske data
| CD154 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
| IgG1 κ |
| Flow Cytometry | |
| TRAP1 | |
| Vol. Per test: 5μL | |
| CD40LG; CD40 ligand; CD40L; TNFSF5; T-BAM; TRAP | |
| 25 Tests | |
| Cell Biology | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Reseach Use Only.
Ser du en mulighed for forbedring?Del en indholdskorrektion