Learn More
PeproTech CD154 (CD40 Ligand) (soluble) Polyclonal Antibody, PeproTech®, Invitrogen™

Goat Polyclonal Antibody
£177.00 - £1468.00
Specifications
| Antigen | CD154 (CD40 Ligand) (soluble) |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17824163
|
PeproTech
500-P142G-50UG |
50 μg |
£177.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17814583
|
PeproTech
500-P142G-100UG |
100 μg |
£228.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17814063
![]() |
PeproTech
500-P142G-1MG |
1 mg |
£1468.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: MQKGDQNPQI AAHVISEASS KTTSVLQWAE KGYYTMSNNL VTLENGKQLT VKRQGLYYIY AQVTFCSNRE ASSQAPFIAS LWLKSPGRFE RILLRAANTH SSAKPCGQQS IHLGGVFELQ PGASVFVNVT DPSQVSHGTG FTSFGLLKL. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human sCD40 Ligand. Anti-Human sCD40 Ligand-specific antibody was purified by affinity chromatography employing an immobilized Human sCD40 Ligand matrix. Sandwich ELISA: To detect Human sCD40 Ligand by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human sCD40 Ligand (500-P142GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human sCD40 Ligand. Western Blot: To detect Human sCD40 Ligand by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human sCD40 Ligand is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.Specifications
| CD154 (CD40 Ligand) (soluble) | |
| ELISA, Western Blot | |
| Unconjugated | |
| Goat | |
| Human | |
| P29965 | |
| 959 | |
| E.coli-derived Recombinant Human sCD40 Ligand. | |
| Antigen affinity chromatography | |
| CD40LG |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5 | |
| CD40LG | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.