missing translation for 'onlineSavingsMsg'
Learn More

CD154 (CD40 Ligand) (soluble) Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

Brand:  Invitrogen 500-P142GBT-1MG

 View more versions of this product

Product Code. 17804383

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: MQKGDQNPQI AAHVISEASS KTTSVLQWAE KGYYTMSNNL VTLENGKQLT VKRQGLYYIY AQVTFCSNRE ASSQAPFIAS LWLKSPGRFE RILLRAANTH SSAKPCGQQS IHLGGVFELQ PGASVFVNVT DPSQVSHGTG FTSFGLLKL. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human sCD40 Ligand. Anti-Human sCD40 Ligand-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human sCD40 Ligand by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human sCD40 Ligand (500-P142G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human sCD40 Ligand. Western Blot: To detect hsCD40L by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hsCD40L is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P142GBT-1MG will be provided as 2 x 500 μg

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD154 (CD40 Ligand) (soluble)
Polyclonal
Biotin
CD40LG
CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5
Goat
Antigen affinity chromatography
RUO
959
-20°C
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P29965
CD40LG
E.coli-derived Recombinant Human sCD40 Ligand
2 x 500 μg
Primary
Human
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.