Learn More
Invitrogen™ CD40L (Soluble) Human Matched Antibody Pair
Matched Antibody Pair
Brand: Invitrogen™ BMS239MST
Includes:
- 1 vial (5.5mL) monoclonal Coating Antibody to human sCD40L (100μg/mL)
- 1 vial (110μL) HRP-Conjugate anti-human sCD40L monoclonal antibody
- 1 vial human sCD40L Standard protein lyophilized, 1μg/mL upon reconstitution
- 2 vials (50mL) Sample Diluent
Description
The Human CD40L Matched Antibody Pair Kit comes with pre-matched antibody pairs,standards,and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human CD40L. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate,which is an overnight process. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
CD154 is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD154 stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD154 has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. Further,CD154 also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma,TNF-alpha,and IL-2. A defect in CD154 gene results in an inability to undergo immunoglobulin class switch and is associated with hyper-IgM syndrome. CD154 is a member of the tumor necrosis factor family,and is expressed primarily on activated CD4+lymphocytes,but also on mast cells,basophils,eosinophils and human dendritic cells. CD154 is also found on some epithelial cells,carcinomas and lymphoid dendritic cells. CD154 also appears to be involved in the transduction of regulatory signals for cellular functions such as B cell proliferation and differentiation.
Specifications
| CD154 (CD40 Ligand), sCD154 (sCD40 Ligand) | |
| Monoclonal | |
| P29965 | |
| CD40LG | |
| RUO | |
| 959 | |
| -20°C |
| ELISA | |
| HRP | |
| TRAP, HIGM 1, T cell antigen Gp39, CD154 | |
| 10 x 96 Tests | |
| Primary | |
| Human | |
| ELISA Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.