Learn More
CD154 (CD40 Ligand) Monoclonal Antibody (24-31), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Specifications
Antigen | CD154 (CD40 Ligand) |
---|---|
Clone | 24-31 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15538496
|
Invitrogen
25-1548-42 |
100 Tests |
N/A
|
N/A | |||||
Description
Description: The 24-31 monoclonal antibody reacts with human CD154, a 39 kDa transmembrane glycoprotein also known as gp39 and CD40 ligand (CD40L). CD154 is a member of the TNF superfamily and is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells including B cells, monocytes/macrophages and dendritic cells, CD154 serves a crucial function in T-APC cognate interaction. CD154 interaction with CD40 transduces signals for T-dependent B-cell activation and induces B cell cycle entry. 24-31 cross-reacts with the cynomolgus monkey CD154. Applications Reported: This 24-31 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 24-31 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD154 (CD40 Ligand) Monoclonal antibody specifically detects CD154 (CD40 Ligand) in Cynomolgus Monkey, Human samples. It is validated for Flow Cytometry.Specifications
CD154 (CD40 Ligand) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P29965 | |
CD40LG | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD40LG |
24-31 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Cynomolgus Monkey, Human | |
102138630, 959 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.