Learn More
CD154 (CD40 Ligand) Monoclonal Antibody (MR1), Biotin, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
£55.75 - £238.00
Specifications
| Antigen | CD154 (CD40 Ligand) |
|---|---|
| Clone | MR1 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15229749
|
Invitrogen
13-1541-81 |
50 μg | |||||||
15239749
![]() |
Invitrogen™
13-1541-82 |
100 μg |
£135.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15249749
|
Invitrogen™
13-1541-85 |
500 μg |
£238.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The MR1 monoclonal antibody reacts with mouse CD154, a 39 kDa transmembrane glycoprotein also known as CD40 ligand (CD40L). CD154 is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells (APC) including B cells, monocytes/macrophages, and dendritic cells, it serves a crucial function in T cell-APC cognate interaction. CD154-interaction with CD40 transduces signals for T-dependent B cell activation and induces B cells to enter the cell cycle. For staining for flow cytometric analysis, it is important to stimulate enriched T cells or enriched CD4 cells (using depletion strategy) prior to staining with MR1. Applications Reported: The MR1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MR1 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.Specifications
| CD154 (CD40 Ligand) | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P27548 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| MR1 | |
| Flow Cytometry | |
| Biotin | |
| Armenian Hamster | |
| Mouse | |
| 21947 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.