missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD154 Armenian Hamster, Biotin, Clone: MR1, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand: EnquireBio QAB88-B-25ug
Additional Details : Weight : 0.01000kg
Specifications
CD154 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Cd40lg | |
Protein A/G Purified | |
RUO | |
21947 | |
IgG |
Flow Cytometry | |
MR1 | |
Biotin | |
NP_035746 | |
Armenian Hamster | |
25 μg | |
Primary | |
Store at 2-4°C. |