missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD151 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595996
This item is not returnable.
View return policy
Description
Immunogen sequence: AYYQQLNTEL KENLKDTMTK RYHQPGHEAV TSAVDQLQQE FHCCGSNNSQ DWRDSEWIRS QEAGGRVVPD SCCKTVVALC GQRDHASNIY KVEGGCITKL ETFIQEHLR; Positive Samples: Mouse skeletal muscle; Cellular Location: Membrane, Multi-pass membrane protein.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It is involved in cellular processes including cell adhesion and may regulate integrin trafficking and/or function. This protein enhances cell motility, invasion and metastasis of cancer cells. Multiple alternatively spliced transcript variants that encode the same protein have been described for this gene.
Specifications
| CD151 | |
| Polyclonal | |
| Unconjugated | |
| CD151 | |
| CD 151; CD151; CD151 antigen; CD151 antigen (Raph blood group); CD151 molecule; CD151 molecule (Raph blood group); GP27; hemidesmosomal tetraspanin CD151; membrane glycoprotein SFA-1; MER2; Peta3; PETA-3; platelet endothelial tetraspan antigen-3; platelet surface glycoprotein gp27; platelet-endothelial cell tetraspan antigen 3; platelet-endothelial tetraspan antigen 3; RAPH; SFA1; SFA-1; Tetraspanin-24; TSPAN24; tspan-24 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12476, 64315, 977 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O35566, P48509, Q9QZA6 | |
| CD151 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 113-221 of human CD151 (NP_0043482). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction