missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD151 Monoclonal Antibody (455807)
Description
This antibody stains mouse CD151 transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It is involved in cellular processes including cell adhesion and may regulate integrin trafficking and/or function. This protein enhances cell motility, invasion and metastasis of cancer cells. Multiple alternatively spliced transcript variants that encode the same protein have been described for this gene.
Specifications
Specifications
| Antigen | CD151 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 455807 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CD151 |
| Gene Accession No. | O35566 |
| Gene Alias | CD 151; CD151; CD151 antigen; CD151 antigen (Raph blood group); CD151 molecule; CD151 molecule (Raph blood group); GP27; hemidesmosomal tetraspanin CD151; membrane glycoprotein SFA-1; MER2; Peta3; PETA-3; platelet endothelial tetraspan antigen-3; platelet surface glycoprotein gp27; platelet-endothelial cell tetraspan antigen 3; platelet-endothelial tetraspan antigen 3; RAPH; SFA1; SFA-1; Tetraspanin-24; TSPAN24; tspan-24 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?