missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD151 Antibody (Pierre Fabre patent anti-CD151) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28136-100ug
This item is not returnable.
View return policy
Description
CD151 Monoclonal antibody specifically detects CD151 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD151 | |
| Monoclonal | |
| Unconjugated | |
| P48509 | |
| Human | |
| Protein A purified | |
| RUO | |
| 977 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Pierre Fabre patent anti-CD151 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
| CD151 | |
| 100 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction