missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD150 (SLAM) Hamster, anti-Mouse, Alexa Fluor 488, Clone: TC15-12F12.2, STEMCELL Technologies
Description
- The TC15-12F12
- 2 antibody reacts with CD150 (signaling lymphocyte activation molecule or SLAM), an ∼75 kDa type I transmembrane glycoprotein that plays multiple roles in the immune response by serving as a cell adhesion molecule and/or coreceptor
- It is differentially expressed by T cells, immature thymocytes, B cells, dendritic cells, macrophages, and endothelial cells
- The expression pattern differs according to cell type and activation status
- Expression is rapidly induced upon activation of T cells, B cells and dendritic cells, with synthesis by T cells being maintained on Th1 but not Th2 clones
- CD150-mediated co-stimulation of TCR-activated T cells enhances the production of IFN-γ by Th1 cells, a response that can be augmented by binding of the TC15-12F12
- 2 antibody
- CD150 is thought to mediate signal transduction by associating with the intracellular protein tyrosine phosphatase, SHP-2
Specifications
Specifications
| Antigen | CD150 (SLAM) |
| Applications | Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | TC15-12F12.2 |
| Conjugate | Alexa Fluor 488 |
| Host Species | Hamster |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Target Species | Mouse |
| Isotype | IgG2a λ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?