missing translation for 'onlineSavingsMsg'
Learn More

CD15/Lewis X Mouse anti-Human, Clone: BRA-4F1, Novus Biologicals™

Product Code. 18359641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
Unit Size:
0.05mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359641 0.025 mg 25µg
18351712 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18359641 Supplier Novus Biologicals Supplier No. NBP3120690.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD15/Lewis X Monoclonal antibody specifically detects CD15/Lewis X in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD15/Lewis X
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone BRA-4F1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias CD 15, CD-15, Lewis X, Lewis-X, LeX, SSEA-1, stage-specific embryonic antigen 1
Host Species Mouse
Immunogen This antibody was raised by immunzing mice with human CD15/Lewis X.
Purification Method Affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2526
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.