missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD148 Syrian Hamster anti-Mouse, Oligo, Clone: 8A-1, BD™ AbSeq ™
Syrian Hamster Monoclonal Antibody
Brand: BD Biosciences 940485
This item is not returnable.
View return policy
Description
CD148 Monoclonal antibody specifically detects CD148 in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD148 | |
| Monoclonal | |
| Oligo | |
| Ptprj; R-PTP-J; BET; Byp; DEP-1; PTPbeta2; Ptpb2; R-PTP-eta; Scc-1; Scc1 | |
| Mouse CD148 Transfected Cell Line | |
| RUO | |
| Mouse | |
| IgG1 |
| Single Cell 3' Sequencing | |
| Clone 8A-1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Syrian Hamster | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction