missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD148 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119081
This item is not returnable.
View return policy
Description
CD148 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
CD148 is a highly glycosylated transmembrane protein of up to 250 kDa, containing eight fibronectin type III domains in the extracellular region and one tyrosine phosphatase domain in the cytoplasmic region. Expression is found broadly on fibroblasts, epithelial cells, lymphocytes (B, T and NK cells), and myeloid cells (monocytes and granulocytes). CD148 can dephosphorylate proteins such as LAT and PLC-gamma, and is involved in the downregulation of T cell activation. Conversely, CD148 expression can be upregulated following T cell activation.
Specifications
| CD148 | |
| Polyclonal | |
| Unconjugated | |
| PTPRJ | |
| AI450271; BET; Byp; CD148; CD148 antigen; colon tumor susceptibility 1; density enhanced phosphatase-1; density-enhanced phosphatase 1; DEP1; DEP-1; HPTP beta-like tyrosine phosphatase; HPTP eta; HPTPeta; human density enhanced phosphatase-1; protein tyrosine phosphatase, receptor type J; protein tyrosine phosphatase, receptor type, J; protein tyrosine phosphatase, receptor type, J polypeptide; protein-tyrosine phosphatase eta; Protein-tyrosine phosphatase receptor type J; Ptpb2; PTPbeta2; Ptprj; Receptor-type tyrosine-protein phosphatase eta; R-PTP-ETA; RPTPJ; R-PTP-J; Scc1; Scc-1; Susceptibility to colon cancer 1; susceptibility to colon cancer 1, mouse, homolog of | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 19271, 5795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q12913, Q64455 | |
| PTPRJ | |
| Recombinant protein Density Enhanced Phosphatase 1. The antigen corresponds to amino acid range 1061-1293 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction