missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD147 Rat anti-Mouse , Alexa Fluor 488, Clone: RL73 (also known as RL73.2), BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 566869
This item is not returnable.
View return policy
Description
CD147 Monoclonal antibody specifically detects CD147 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD147 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Bsg; Basigin; BASI; EMMPRIN; gp 42; HT7; HT-7; Neurothelin | |
| Mouse EL4 Cell Subline | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| RL73 (also known as RL73.2) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction