missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD146 Recombinant Rabbit Monoclonal Antibody (241), FITC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£113.00 - £283.00
Specifications
| Antigen | CD146 |
|---|---|
| Clone | 241 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019630
![]() |
Invitrogen™
MA552424 |
100 Tests |
£283.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019351
|
Invitrogen™
MA552425 |
25 Tests |
£113.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD146 Recombinant Monoclonal antibody specifically detects CD146 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD146 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.03% ProClin 300 | |
| 1-gicerin; AV025631; CD146; CD149; Cell surface glycoprotein MUC18; cell surface glycoprotein P1H12; Gicerin; HEMCAM; l-gicerin; l-gicerin precursor; l-gicerin protein; LOW QUALITY PROTEIN: cell surface glycoprotein MUC18; MCAM; melanoma adhesion molecule; melanoma cell adhesion molecule; Melanoma-associated antigen A32; melanoma-associated antigen MUC18; Muc18; mucin-18; s-endo; S-endo 1 endothelial-associated antigen; s-gicerin; s-gicerin precursor | |
| Recombinant Mouse CD146/MCAM Protein (Val 24-Val563). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| MCAM |
| 241 | |
| Flow Cytometry | |
| FITC | |
| Rabbit | |
| Mouse | |
| Q8R2Y2 | |
| 84004 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title