missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD142 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527278
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII. The resulting complex provides a catalytic event that is responsible for initiation of the coagulation protease cascades by specific limited proteolysis. Unlike the other cofactors of these protease cascades, which circulate as nonfunctional precursors, this factor is a potent initiator that is fully functional when expressed on cell surfaces. There are 3 distinct domains of this factor: extracellular, transmembrane, and cytoplasmic. This protein is the only one in the coagulation pathway for which a congenital deficiency has not been described.
Specifications
| CD142 | |
| Polyclonal | |
| Unconjugated | |
| F3 | |
| AA409063; CD142; Cf3; Cf-3; Coagulation factor III; coagulation factor III (thromboplastin, tissue factor); coagulation factor III, tissue factor; F3; TF; TFA; Thromboplastin; Tissue factor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2152 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P13726 | |
| F3 | |
| Recombinant protein encompassing a sequence within the center region of human Coagulation factor III/Tissue Factor. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction