missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD142 Monoclonal Antibody (K06319_4H12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556353
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: LTDEIVKDVK QTYLARVFSY PAGNVESTGS AGEPLYENSP EFTPYLETNL GQPTIQSFEQ VGTKVNVTVE
This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII. The resulting complex provides a catalytic event that is responsible for initiation of the coagulation protease cascades by specific limited proteolysis. Unlike the other cofactors of these protease cascades, which circulate as nonfunctional precursors, this factor is a potent initiator that is fully functional when expressed on cell surfaces. There are 3 distinct domains of this factor: extracellular, transmembrane, and cytoplasmic. This protein is the only one in the coagulation pathway for which a congenital deficiency has not been described.
Specifications
| CD142 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P13726 | |
| F3 | |
| Recombinant human F3 (immunogen range is 91-160). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| K06319_4H12 | |
| Unconjugated | |
| F3 | |
| AA409063; CD142; Cf3; Cf-3; Coagulation factor III; coagulation factor III (thromboplastin, tissue factor); coagulation factor III, tissue factor; F3; TF; TFA; Thromboplastin; Tissue factor | |
| Mouse | |
| Protein A | |
| RUO | |
| 2152 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction