missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD140b (PDGFRB) Monoclonal Antibody (APB5), Super Bright™ 600, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£129.00 - £367.00
Specifications
| Antigen | CD140b (PDGFRB) |
|---|---|
| Clone | APB5 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
Rat Monoclonal Antibody
PDGFRB is a receptor tyrosine kinase and has been implicated in atherogenesis and in cell transformation by the SIS oncogene. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. The gene is flanked on chromosome 5 by the genes for granulocyte-macrophage colony-stimulating factor and macrophage-colony stimulating factor receptor; all three genes may be implicated in the 5-q syndrome. A translocation between chromosomes 5 and 12, that fuses PDGFRB (CD140b) to that of the translocation, ETV6, leukemia gene, results in chronic myeloproliferative disorder with eosinophilia.Specifications
| CD140b (PDGFRB) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 18596 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| APB5 | |
| Flow Cytometry | |
| Super Bright 600 | |
| Rat | |
| Mouse | |
| P05622 | |
| Pdgfrb | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Pdgfrb |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title