Learn More
CD14 Monoclonal Antibody (61D3), PerCP-Cyanine5.5, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 45-0149-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The 61D3 monoclonal antibody reacts with human CD14, a 53-55 kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14. Applications Reported: This 61D3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 61D3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P08571 | |
Cd14 | |
Affinity chromatography | |
RUO | |
929 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
61D3 | |
PerCP-Cyanine5.5 | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.